accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHNFFFCl

CAS RN 76041-71-9

IUPAC Name: 3-chloro-5-(trifluoromethyl)-1,2-dihydropyridin-2-one
Synonyms: 3-chloro-5-(trifluoromethyl)-1H-pyridin-2-one
Molecular Weight (g/mol): 197.54
Molecular Formula: C6H3ClF3NO
InChi Key: AJPOOWWMZOPUCG-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=CNC(=O)C(Cl)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results