accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 76095-16-4

IUPAC Name: (2S)-1-[(2S)-2-{[(2R)-1-ethoxy-1-oxo-4-phenylbutan-2-yl]amino}propanoyl]pyrrolidine-2-carboxylic acid; (2Z)-but-2-enedioic acid
Synonyms: maleic acid; vasotec
Molecular Weight (g/mol): 492.53
Molecular Formula: C24H32N2O9
InChi Key: OYFJQPXVCSSHAI-BMSLDGIRSA-N
SMILES: OC(=O)\C=C/C(O)=O.CCOC(=O)[C@@H](CCC1=CC=CC=C1)N[C@@H](C)C(=O)N1CCC[C@H]1C(O)=O
Percent Purity 
  • (1)
  • (2)
Grade 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)