CAS RN 761-01-3
IUPAC Name:
sulfonylideneoxidane; triethylamine
Synonyms:
sulfur trioxide; triethylamine
Molecular Weight (g/mol):
181.25
Molecular Formula:
C6H15NO3S
InChi Key:
YYHPEVZFVMVUNJ-UHFFFAOYSA-N
SMILES:
O=S(=O)=O.CCN(CC)CC