accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClNOONClClCl

CAS RN 76150-91-9

IUPAC Name: 3,5-dichloro-2-{4-[(3,5-dichloropyridin-2-yl)oxy]phenoxy}pyridine
Synonyms: 3,5-dichloro-2-{4-[(3,5-dichloropyridin-2-yl)oxy]phenoxy}pyridine
Molecular Weight (g/mol): 402.05
Molecular Formula: C16H8Cl4N2O2
InChi Key: BAFKRPOFIYPKBQ-UHFFFAOYSA-N
SMILES: ClC1=CN=C(OC2=CC=C(OC3=NC=C(Cl)C=C3Cl)C=C2)C(Cl)=C1
Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results