accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3CSSHN

CAS RN 76186-04-4

IUPAC Name: 4-(propan-2-yl)-1,3-thiazolidine-2-thione
Synonyms: 4-isopropyl-1,3-thiazolidine-2-thione
Molecular Weight (g/mol): 161.28
Molecular Formula: C6H11NS2
InChi Key: CWIZUGZKLJDJLE-UHFFFAOYNA-N
SMILES: CC(C)C1CSC(=S)N1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results