accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 762-29-8

IUPAC Name: (5E,9E)-6,10,14-trimethylpentadeca-5,9,13-trien-2-one
Synonyms: trimethylpentadecatrien-2-one
Molecular Weight (g/mol): 262.44
Molecular Formula: C18H30O
InChi Key: LTUMRKDLVGQMJU-IUBLYSDUSA-N
SMILES: CC(C)=CCC\C(C)=C\CC\C(C)=C\CCC(C)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results