accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 762-62-9

IUPAC Name: 4,4-dimethylpent-1-ene
Synonyms: 1-pentene, 4,4-dimethyl-
Molecular Weight (g/mol): 98.19
Molecular Formula: C7H14
InChi Key: KLCNJIQZXOQYTE-UHFFFAOYSA-N
SMILES: CC(C)(C)CC=C
Boiling Point 
  • (3)
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (4)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (2)
  • (3)
  • (3)
  • (2)
  • (1)
  • (2)
  • (4)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)