accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CClClO

CAS RN 7623-09-8

IUPAC Name: 2-chloropropanoyl chloride
Synonyms: 2-chloropropanoyl chloride
Molecular Weight (g/mol): 126.96
Molecular Formula: C3H4Cl2O
InChi Key: JEQDSBVHLKBEIZ-UHFFFAOYNA-N
SMILES: CC(Cl)C(Cl)=O
Boiling Point 
  • (3)
  • (3)
  • (3)
Molecular Weight (g/mol) 
  • (6)
  • (3)
Percent Purity 
  • (3)
  • (3)
  • (3)
Quantity 
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (3)
  • (6)
  • (3)
  • (3)
  • (3)
  • (6)
  • (3)
  • (3)
  • (3)
  • (3)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (6)