accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 76265-70-8

IUPAC Name: (2S)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-4-methanesulfinylbutanoic acid
Synonyms: (2S)-2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}-4-methanesulfinylbutanoic acid|FMOC-MET(O)-OH
Molecular Weight (g/mol): 387.45
Molecular Formula: C20H21NO5S
InChi Key: CEHRSUBRZOGRSW-HSYKDVHTSA-N
SMILES: CS(=O)CC[C@H](NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)