accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 763-30-4

IUPAC Name: 2-methylpenta-1,4-diene
Synonyms: 2-methyl-1,4-pentadiene
Molecular Weight (g/mol): 82.15
Molecular Formula: C6H10
InChi Key: DRWYRROCDFQZQF-UHFFFAOYSA-N
SMILES: CC(=C)CC=C
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results