accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 763109-71-3

IUPAC Name: 5-(thiophen-2-yl)-1,2-oxazole-3-carboxylic acid
Synonyms: 5-(thiophen-2-yl)-1,2-oxazole-3-carboxylic acid
Molecular Weight (g/mol): 195.19
Molecular Formula: C8H5NO3S
InChi Key: OBTRHBQUXRJHJR-UHFFFAOYSA-N
SMILES: OC(=O)C1=NOC(=C1)C1=CC=CS1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results