accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 764-48-7

HOOCH2

CAS RN 764-48-7

IUPAC Name: 2-(ethenyloxy)ethan-1-ol
Synonyms: ethanol, 2-(ethenyloxy)-
Molecular Weight (g/mol): 88.11
Molecular Formula: C4H8O2
InChi Key: VUIWJRYTWUGOOF-UHFFFAOYSA-N
SMILES: OCCOC=C
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results