accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COHOHO

CAS RN 764-67-0

IUPAC Name: 2-hydroxyhexadecanoic acid
Synonyms: 2-hydroxy palmitic acid
Molecular Weight (g/mol): 272.43
Molecular Formula: C16H32O3
InChi Key: JGHSBPIZNUXPLA-UHFFFAOYNA-N
SMILES: CCCCCCCCCCCCCCC(O)C(O)=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results