accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 764-93-2

IUPAC Name: dec-1-yne
Synonyms: 1-decyne
Molecular Weight (g/mol): 138.25
Molecular Formula: C10H18
InChi Key: ILLHQJIJCRNRCJ-UHFFFAOYSA-N
SMILES: CCCCCCCCC#C
Boiling Point 
  • (2)
  • (1)
  • (3)
  • (2)
Molecular Weight (g/mol) 
  • (3)
  • (5)
Percent Purity 
  • (2)
  • (6)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (1)
  • (2)
  • (5)
  • (2)
  • (1)
  • (3)
  • (2)
  • (3)
  • (5)
  • (2)
  • (6)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (4)