CAS RN 7640-33-7
IUPAC Name:
7-bromo-5-chloroquinolin-8-ol
Synonyms:
7-bromo-5-chloroquinolin-8-ol
Molecular Weight (g/mol):
258.5
Molecular Formula:
C9H5BrClNO
InChi Key:
ADJQQSUBKRASQN-UHFFFAOYSA-N
SMILES:
OC1=C2N=CC=CC2=C(Cl)C=C1Br
1
–
1
of
1
results
1
7-Bromo-5-chloro-8-hydroxyquinoline 98.0+%, TCI America™
CAS: 7640-33-7 Molecular Formula: C9H5BrClNO Molecular Weight (g/mol): 258.499 MDL Number: MFCD00044601 InChI Key: ADJQQSUBKRASQN-UHFFFAOYSA-N PubChem CID: 82095 IUPAC Name: 7-bromo-5-chloroquinolin-8-ol SMILES: C1=CC2=C(C(=C(C=C2Cl)Br)O)N=C1