accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 7646-75-5

CH3SiH3CONa

CAS RN 7646-75-5

IUPAC Name: sodium dimethyl(phenyl)silanolate
Synonyms: sodium dimethyl(phenyl)silanolate
Molecular Weight (g/mol): 174.25
Molecular Formula: C8H11NaOSi
InChi Key: NPZJKOMUCXNLBN-UHFFFAOYSA-N
SMILES: [Na+].C[Si](C)([O-])C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results