accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OON

CAS RN 76469-88-0

IUPAC Name: methyl 4-(cyanomethyl)benzoate
Synonyms: methyl 4-(cyanomethyl)benzoate
Molecular Weight (g/mol): 175.19
Molecular Formula: C10H9NO2
InChi Key: XRZGMNGGCZTNGE-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC=C(CC#N)C=C1
Boiling Point 
  • (1)
  • (2)
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)