CAS RN 7647-10-1
Palladium(II) chloride, 59% Pd
CAS: 7647-10-1 Molecular Formula: Cl2Pd Molecular Weight (g/mol): 177.32 MDL Number: MFCD00003558 InChI Key: PIBWKRNGBLPSSY-UHFFFAOYSA-L Synonym: palladium chloride,palladium ii chloride,palladium dichloride,palladous chloride,pdcl2,palladium 2+ chloride,palladium chloride pdcl2,enplate activator 440 PubChem CID: 24290 ChEBI: CHEBI:53434 SMILES: [Cl-].[Cl-].[Pd++]
Palladium(II) chloride, 99.9% (metals basis), Pd 59.0% min
CAS: 7647-10-1 Molecular Formula: Cl2Pd Molecular Weight (g/mol): 177.32 MDL Number: MFCD00003558 InChI Key: PIBWKRNGBLPSSY-UHFFFAOYSA-L PubChem CID: 24290 ChEBI: CHEBI:53434 SMILES: [Cl-].[Cl-].[Pd++]
Palladium(II) chloride, Premion™, 99.999% (metals basis), Pd 59.5% min
CAS: 7647-10-1 Molecular Formula: Cl2Pd Molecular Weight (g/mol): 177.32 MDL Number: MFCD00003558 InChI Key: PIBWKRNGBLPSSY-UHFFFAOYSA-L Synonym: palladium chloride,palladium ii chloride,palladium dichloride,palladous chloride,pdcl2,palladium 2+ chloride,palladium chloride pdcl2,enplate activator 440 PubChem CID: 24290 ChEBI: CHEBI:53434 SMILES: [Cl-].[Cl-].[Pd++]
Hydrochloric Acid, 5.0N Solution, Biotech Reagent, J.T. Baker™
CAS: 7647-10-1 Molecular Formula: Cl2Pd Molecular Weight (g/mol): 177.32 MDL Number: MFCD00003558 InChI Key: PIBWKRNGBLPSSY-UHFFFAOYSA-L Synonym: palladium chloride,palladium ii chloride,palladium dichloride,palladous chloride,pdcl2,palladium 2+ chloride,palladium chloride pdcl2,enplate activator 440 PubChem CID: 24290 ChEBI: CHEBI:53434 IUPAC Name: palladium(2+) dichloride SMILES: [Cl-].[Cl-].[Pd++]
Palladium(II) chloride, 99.999%, (trace metal basis)
CAS: 7647-10-1 Molecular Formula: Cl2Pd Molecular Weight (g/mol): 177.32 MDL Number: MFCD00003558 InChI Key: PIBWKRNGBLPSSY-UHFFFAOYSA-L Synonym: palladium chloride,palladium ii chloride,palladium dichloride,palladous chloride,pdcl2,palladium 2+ chloride,palladium chloride pdcl2,enplate activator 440 PubChem CID: 24290 ChEBI: CHEBI:53434 SMILES: [Cl-].[Cl-].[Pd++]
Palladium(II) chloride, solution, Pd 9.0-11.0% w/w (cont. Pd)
CAS: 7647-10-1 Molecular Formula: Cl2Pd Molecular Weight (g/mol): 177.32 MDL Number: MFCD00003558 InChI Key: PIBWKRNGBLPSSY-UHFFFAOYSA-L Synonym: palladium chloride,palladium ii chloride,palladium dichloride,palladous chloride,pdcl2,palladium 2+ chloride,palladium chloride pdcl2,enplate activator 440 PubChem CID: 24290 ChEBI: CHEBI:53434 IUPAC Name: dichloropalladium SMILES: [Cl-].[Cl-].[Pd++]
Palladium(II) Chloride 98.0+%, TCI America™
CAS: 7647-10-1 Molecular Formula: Cl2Pd Molecular Weight (g/mol): 177.32 MDL Number: MFCD00003558 InChI Key: PIBWKRNGBLPSSY-UHFFFAOYSA-L Synonym: palladium chloride,palladium ii chloride,palladium dichloride,palladous chloride,pdcl2,palladium 2+ chloride,palladium chloride pdcl2,enplate activator 440 PubChem CID: 24290 ChEBI: CHEBI:53434 IUPAC Name: palladium(2+) dichloride SMILES: [Cl-].[Cl-].[Pd++]
Hydrochloric Acid Solution, 2.5 N, ENG Scientific
CAS: 7647-10-1 Molecular Formula: Cl2Pd Molecular Weight (g/mol): 177.32 MDL Number: MFCD00003558 InChI Key: PIBWKRNGBLPSSY-UHFFFAOYSA-L Synonym: palladium chloride,palladium ii chloride,palladium dichloride,palladous chloride,pdcl2,palladium 2+ chloride,palladium chloride pdcl2,enplate activator 440 PubChem CID: 24290 ChEBI: CHEBI:53434 IUPAC Name: palladium(2+) dichloride SMILES: [Cl-].[Cl-].[Pd++]
Sigma Aldrich Palladium(II) Chloride
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
Sigma Aldrich Palladium(II) Chloride
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
Sigma Aldrich Palladium(II) Chloride
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
Sigma Aldrich Palladium(II) Chloride
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
Hydrochloric acid, Acculute Std. Volumetric Soln., Final Concentration 1.0N, Thermo Scientific™
CAS: 7647-10-1 Molecular Formula: Cl2Pd Molecular Weight (g/mol): 177.32 MDL Number: MFCD00003558 InChI Key: PIBWKRNGBLPSSY-UHFFFAOYSA-L PubChem CID: 24290 ChEBI: CHEBI:53434 SMILES: [Cl-].[Cl-].[Pd++]
Palladium(II) chloride, solution, Pd 20-25% w/w (cont. Pd), Thermo Scientific™
CAS: 7647-10-1 Molecular Formula: Cl2Pd Molecular Weight (g/mol): 177.32 MDL Number: MFCD00003558 InChI Key: PIBWKRNGBLPSSY-UHFFFAOYSA-L Synonym: palladium chloride,palladium ii chloride,palladium dichloride,palladous chloride,pdcl2,palladium 2+ chloride,palladium chloride pdcl2,enplate activator 440 PubChem CID: 24290 ChEBI: CHEBI:53434 IUPAC Name: dichloropalladium SMILES: [Cl-].[Cl-].[Pd++]