CAS RN 7647-15-6
Sodium Bromide (Granular/Certified ACS), Fisher Chemical™
CAS: 7647-15-6 Molecular Formula: BrNa Molecular Weight (g/mol): 102.89 MDL Number: MFCD00003475 InChI Key: JHJLBTNAGRQEKS-UHFFFAOYSA-M Synonym: sodium bromide,bromide salt of sodium,sodium bromide nabr,sedoneural,sodiumbromide,trisodium tribromide,bromnatrium,nabr,bromnatrium german,caswell no. 750a PubChem CID: 253881 ChEBI: CHEBI:63004 IUPAC Name: sodium bromide SMILES: [Na+].[Br-]
Bromide Standard, 1000 ppm Br-, Ricca Chemical
CAS: 7647-15-6 Molecular Formula: BrNa Molecular Weight (g/mol): 102.89 MDL Number: MFCD00003475 InChI Key: JHJLBTNAGRQEKS-UHFFFAOYSA-M PubChem CID: 253881 ChEBI: CHEBI:63004 IUPAC Name: sodium bromide SMILES: [Na+].[Br-]
Sodium bromide, ACS reagent
CAS: 7647-15-6 Molecular Formula: BrNa Molecular Weight (g/mol): 102.89 MDL Number: MFCD00003475 InChI Key: JHJLBTNAGRQEKS-UHFFFAOYSA-M Synonym: sodium bromide,bromide salt of sodium,sodium bromide nabr,sedoneural,sodiumbromide,trisodium tribromide,bromnatrium,nabr,bromnatrium german,caswell no. 750a PubChem CID: 253881 ChEBI: CHEBI:63004 SMILES: [Na+].[Br-]
Sodium bromide, 99+% (dry wt.), water <1.0%
CAS: 7647-15-6 Molecular Formula: BrNa Molecular Weight (g/mol): 102.89 MDL Number: MFCD00003475 InChI Key: JHJLBTNAGRQEKS-UHFFFAOYSA-M Synonym: sodium bromide,bromide salt of sodium,sodium bromide nabr,sedoneural,sodiumbromide,trisodium tribromide,bromnatrium,nabr,bromnatrium german,caswell no. 750a PubChem CID: 253881 ChEBI: CHEBI:63004 SMILES: [Na+].[Br-]
Bromide Standard, 1000 ppm Br-, Fisherbrand
CAS: 7647-15-6 Molecular Formula: BrNa Molecular Weight (g/mol): 102.89 MDL Number: MFCD00003475 InChI Key: JHJLBTNAGRQEKS-UHFFFAOYSA-M PubChem CID: 253881 ChEBI: CHEBI:63004 IUPAC Name: sodium bromide SMILES: [Na+].[Br-]
Sodium bromide, 99.5%, for analysis
CAS: 7647-15-6 Molecular Formula: BrNa Molecular Weight (g/mol): 102.89 MDL Number: MFCD00003475 InChI Key: JHJLBTNAGRQEKS-UHFFFAOYSA-M Synonym: sodium bromide,bromide salt of sodium,sodium bromide nabr,sedoneural,sodiumbromide,trisodium tribromide,bromnatrium,nabr,bromnatrium german,caswell no. 750a PubChem CID: 253881 ChEBI: CHEBI:63004 SMILES: [Na+].[Br-]
Bromide Standard, 100 ppm Br-, Ricca Chemical
CAS: 7647-15-6 Molecular Formula: BrNa Molecular Weight (g/mol): 102.89 MDL Number: MFCD00003475 InChI Key: JHJLBTNAGRQEKS-UHFFFAOYSA-M PubChem CID: 253881 ChEBI: CHEBI:63004 IUPAC Name: sodium bromide SMILES: [Na+].[Br-]
Sodium bromide, 99+%, extra pure, anhydrous
CAS: 7647-15-6 Molecular Formula: BrNa Molecular Weight (g/mol): 102.89 MDL Number: MFCD00003475 InChI Key: JHJLBTNAGRQEKS-UHFFFAOYSA-M Synonym: sodium bromide,bromide salt of sodium,sodium bromide nabr,sedoneural,sodiumbromide,trisodium tribromide,bromnatrium,nabr,bromnatrium german,caswell no. 750a PubChem CID: 253881 ChEBI: CHEBI:63004 IUPAC Name: sodium bromide SMILES: [Na+].[Br-]
Sodium bromide, ACS, 99.0% min
CAS: 7647-15-6 Molecular Formula: BrNa Molecular Weight (g/mol): 102.89 MDL Number: MFCD00003475 InChI Key: JHJLBTNAGRQEKS-UHFFFAOYSA-M Synonym: sodium bromide,bromide salt of sodium,sodium bromide nabr,sedoneural,sodiumbromide,trisodium tribromide,bromnatrium,nabr,bromnatrium german,caswell no. 750a PubChem CID: 253881 ChEBI: CHEBI:63004 SMILES: [Na+].[Br-]
Sodium Bromide, Crystal, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™
CAS: 7647-15-6 Molecular Formula: BrNa Molecular Weight (g/mol): 102.89 MDL Number: MFCD00003475 InChI Key: JHJLBTNAGRQEKS-UHFFFAOYSA-M Synonym: sodium bromide,bromide salt of sodium,sodium bromide nabr,sedoneural,sodiumbromide,trisodium tribromide,bromnatrium,nabr,bromnatrium german,caswell no. 750a PubChem CID: 253881 ChEBI: CHEBI:63004 SMILES: [Na+].[Br-]
Sodium Bromide, Granular, ACS, 99%, Spectrum™ Chemical
CAS: 7647-15-6 Molecular Formula: BrNa Molecular Weight (g/mol): 102.89 MDL Number: MFCD00003475 InChI Key: JHJLBTNAGRQEKS-UHFFFAOYSA-M IUPAC Name: sodium bromide SMILES: [Na+].[Br-]
Sodium Bromide, Granular, Purified, 98%, Spectrum™ Chemical
CAS: 7647-15-6 Molecular Formula: BrNa Molecular Weight (g/mol): 102.89 MDL Number: MFCD00003475 InChI Key: JHJLBTNAGRQEKS-UHFFFAOYSA-M IUPAC Name: sodium bromide SMILES: [Na+].[Br-]
Sodium bromide, Puratronic™, 99.9955% (metals basis)
CAS: 7647-15-6 Molecular Formula: BrNa Molecular Weight (g/mol): 102.89 MDL Number: MFCD00003475 InChI Key: JHJLBTNAGRQEKS-UHFFFAOYSA-M Synonym: sodium bromide,bromide salt of sodium,sodium bromide nabr,sedoneural,sodiumbromide,trisodium tribromide,bromnatrium,nabr,bromnatrium german,caswell no. 750a PubChem CID: 253881 ChEBI: CHEBI:63004 SMILES: [Na+].[Br-]
Sodium Bromide, 99.0% min., ACS, MilliporeSigma™
CAS: 7647-15-6 Molecular Formula: BrNa Molecular Weight (g/mol): 102.89 MDL Number: MFCD00003475 InChI Key: JHJLBTNAGRQEKS-UHFFFAOYSA-M Synonym: sodium bromide,bromide salt of sodium,sodium bromide nabr,sedoneural,sodiumbromide,trisodium tribromide,bromnatrium,nabr,bromnatrium german,caswell no. 750a PubChem CID: 253881 ChEBI: CHEBI:63004 IUPAC Name: sodium bromide SMILES: [Na+].[Br-]
Sodium bromide, 97%
CAS: 7647-15-6 Molecular Formula: BrNa Molecular Weight (g/mol): 102.89 MDL Number: MFCD00003475 InChI Key: JHJLBTNAGRQEKS-UHFFFAOYSA-M Synonym: sodium bromide,bromide salt of sodium,sodium bromide nabr,sedoneural,sodiumbromide,trisodium tribromide,bromnatrium,nabr,bromnatrium german,caswell no. 750a PubChem CID: 253881 ChEBI: CHEBI:63004 IUPAC Name: sodium bromide SMILES: [Na+].[Br-]