accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH

CAS RN 765-10-6

IUPAC Name: tetradec-1-yne
Synonyms: tetradec-1-yne
Molecular Weight (g/mol): 194.36
Molecular Formula: C14H26
InChi Key: DZEFNRWGWQDGTR-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCC#C
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)