accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 765-87-7

IUPAC Name: cyclohexane-1,2-dione
Synonyms: 1,2-cyclohexanedione
Molecular Weight (g/mol): 112.13
Molecular Formula: C6H8O2
InChi Key: OILAIQUEIWYQPH-UHFFFAOYSA-N
SMILES: O=C1CCCCC1=O
Boiling Point 
  • (2)
  • (2)
  • (3)
  • (2)
Molecular Weight (g/mol) 
  • (4)
  • (5)
Percent Purity 
  • (2)
  • (2)
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (3)
  • (1)
Physical Form 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (2)
  • (5)
  • (2)
  • (2)
  • (3)
  • (2)
  • (4)
  • (5)
  • (2)
  • (2)
  • (2)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)
  • (3)
  • (1)
  • (2)
  • (3)