accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CP

CAS RN 7650-84-2

IUPAC Name: diphenyl(propyl)phosphane
Synonyms: phosphine, diphenylpropyl-
Molecular Weight (g/mol): 228.28
Molecular Formula: C15H17P
InChi Key: AAXGWYDSLJUQLN-UHFFFAOYSA-N
SMILES: CCCP(C1=CC=CC=C1)C1=CC=CC=C1
Boiling Point 
  • (3)
  • (1)
Percent Purity 
  • (1)
  • (3)
Quantity 
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (3)
  • (3)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)