accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COPOOH3COClPOOCH3OCH3

CAS RN 76541-72-5

IUPAC Name: dimethyl [(4-chlorophenyl)[(dimethoxyphosphoryl)oxy]methyl]phosphonate
Synonyms: mifobate clenicor mifobato mifobatum
Molecular Weight (g/mol): 358.65
Molecular Formula: C11H17ClO7P2
InChi Key: VQHUQHAPWMNBLP-UHFFFAOYNA-N
SMILES: COP(=O)(OC)OC(C1=CC=C(Cl)C=C1)P(=O)(OC)OC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results