accessibility menu, dialog, popup

UserName

Viewing profile as user:
NaH3CCH3OHOH3CH3COO

CAS RN 76584-70-8

IUPAC Name: sodium 2-propylpentanoic acid 2-propylpentanoate
Synonyms: divalproex sodium depakote depakote ER valproate semisodium depakote sprinkle depakote CP sodium divalproate semisodium valproate
Molecular Weight (g/mol): 310.41
Molecular Formula: C16H31NaO4
InChi Key: MSRILKIQRXUYCT-UHFFFAOYSA-M
SMILES: [Na+].CCCC(CCC)C(O)=O.CCCC(CCC)C([O-])=O
Quantity 
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
Grade 
  • (5)
Percent Purity 
  • (5)
Physical Form 
  • (5)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (5)
  • (5)
  • (2)
  • (4)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (5)
  • (5)
  • (5)