accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3OOSH

CAS RN 7659-86-1

IUPAC Name: 2-ethylhexyl 2-sulfanylacetate
Synonyms: 2-ethylhexyl 2-sulfanylacetate
Molecular Weight (g/mol): 204.33
Molecular Formula: C10H20O2S
InChi Key: OWHSTLLOZWTNTQ-UHFFFAOYNA-N
SMILES: CCCCC(CC)COC(=O)CS
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results