accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 765916-91-4

IUPAC Name: 2-fluoro-6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzonitrile
Synonyms: 2-fluoro-6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzonitrile
Molecular Weight (g/mol): 247.08
Molecular Formula: C13H15BFNO2
InChi Key: TXGKJULMHJEZTA-UHFFFAOYSA-N
SMILES: CC1(C)OB(OC1(C)C)C1=C(C#N)C(F)=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results