accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 766-83-6

IUPAC Name: 1-chloro-3-ethynylbenzene
Synonyms: 1-chloro-3-ethynylbenzene
Molecular Weight (g/mol): 136.58
Molecular Formula: C8H5Cl
InChi Key: GRBJPHPMYOUMJV-UHFFFAOYSA-N
SMILES: ClC1=CC=CC(=C1)C#C
Quantity 
  • (2)
  • (1)
Boiling Point 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results