accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 766-92-7

IUPAC Name: [(methylsulfanyl)methyl]benzene
Synonyms: benzene, (methylthio)methyl -
Molecular Weight (g/mol): 138.23
Molecular Formula: C8H10S
InChi Key: OFQPKKGMNWASPN-UHFFFAOYSA-N
SMILES: CSCC1=CC=CC=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results