accessibility menu, dialog, popup

UserName

Viewing profile as user:
BrCH

CAS RN 766-96-1

IUPAC Name: 1-bromo-4-ethynylbenzene
Synonyms: 1-bromo-4-ethynylbenzene
Molecular Weight (g/mol): 181.03
Molecular Formula: C8H5Br
InChi Key: LTLVZQZDXQWLHU-UHFFFAOYSA-N
SMILES: BrC1=CC=C(C=C1)C#C
Molecular Weight (g/mol) 
  • (2)
  • (4)
Percent Purity 
  • (2)
  • (3)
  • (1)
Quantity 
  • (3)
  • (2)
  • (1)
Boiling Point 
  • (1)
  • (2)
  • (2)
Physical Form 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (2)
  • (3)
  • (2)
  • (4)
  • (2)
  • (3)
  • (1)
  • (3)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)