accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3(R)OHONHOOHN

CAS RN 7662-58-0

IUPAC Name: (2R)-2-{[(benzyloxy)carbonyl]amino}-4-methylpentanoic acid; N-cyclohexylcyclohexanamine
Synonyms: (2R)-2-{[(benzyloxy)carbonyl]amino}-4-methylpentanoic acid; dicha
Molecular Weight (g/mol): 446.63
Molecular Formula: C26H42N2O4
InChi Key: FOULZFSGIVQTHX-HVPILOLGNA-N
SMILES: C1CCC(CC1)NC1CCCCC1.CC(C)C[C@@H](NC(=O)OCC1=CC=CC=C1)C(O)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)