accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3SOHN

CAS RN 76632-23-0

IUPAC Name: (2-methyl-1,3-thiazol-4-yl)methanol
Synonyms: (2-methyl-1,3-thiazol-4-yl)methanol
Molecular Weight (g/mol): 129.18
Molecular Formula: C5H7NOS
InChi Key: KRCIFTBSQKDYMH-UHFFFAOYSA-N
SMILES: CC1=NC(CO)=CS1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results