CAS RN 7664-66-6
IUPAC Name:
4-(propan-2-yloxy)aniline
Synonyms:
4-isopropoxyaniline
Molecular Weight (g/mol):
151.21
Molecular Formula:
C9H13NO
InChi Key:
MLNFMFAMNBGAQT-UHFFFAOYSA-N
SMILES:
CC(C)OC1=CC=C(N)C=C1
1
–
1
of
1
results
1
4-Isopropoxyaniline 98.0+%, TCI America™
CAS: 7664-66-6 Molecular Formula: C9H13NO Molecular Weight (g/mol): 151.209 MDL Number: MFCD00059188 InChI Key: MLNFMFAMNBGAQT-UHFFFAOYSA-N PubChem CID: 82112 IUPAC Name: 4-propan-2-yloxyaniline SMILES: CC(C)OC1=CC=C(C=C1)N