CAS RN 7664-93-9
Sulfuric Acid Solution (0.1N (N/10)/Certified), Fisher Chemical™
CAS: 7664-93-9 Molecular Formula: H2O4S Molecular Weight (g/mol): 98.07 MDL Number: MFCD00064589 InChI Key: QAOWNCQODCNURD-UHFFFAOYSA-N Synonym: oil of vitriol,sulphuric acid,dihydrogen sulfate,mattling acid,battery acid,dipping acid,acide sulfurique,electrolyte acid,acidum sulfuricum,vitriol brown oil PubChem CID: 1118 ChEBI: CHEBI:26836 IUPAC Name: sulfuric acid SMILES: OS(O)(=O)=O
Sulfuric Acid Solution (0.2N (N/5)/Certified), Fisher Chemical
CAS: 7664-93-9 Molecular Formula: H2O4S Molecular Weight (g/mol): 98.07 MDL Number: MFCD00064589 InChI Key: QAOWNCQODCNURD-UHFFFAOYSA-N Synonym: oil of vitriol,sulphuric acid,dihydrogen sulfate,mattling acid,battery acid,dipping acid,acide sulfurique,electrolyte acid,acidum sulfuricum,vitriol brown oil PubChem CID: 1118 ChEBI: CHEBI:26836 IUPAC Name: sulfuric acid SMILES: OS(O)(=O)=O
Sulfuric Acid Solution (0.02N (N/50)/Certified), Fisher Chemical™
CAS: 7664-93-9 Molecular Formula: H2O4S Molecular Weight (g/mol): 98.07 MDL Number: MFCD00064589 InChI Key: QAOWNCQODCNURD-UHFFFAOYSA-N Synonym: oil of vitriol,sulphuric acid,dihydrogen sulfate,mattling acid,battery acid,dipping acid,acide sulfurique,electrolyte acid,acidum sulfuricum,vitriol brown oil PubChem CID: 1118 ChEBI: CHEBI:26836 IUPAC Name: sulfuric acid SMILES: OS(O)(=O)=O
Sulfuric Acid, 1N Volumetric Solution, BAKER ANALYZED™ Reagent, J.T. Baker™
CAS: 7664-93-9 Molecular Formula: H2O4S Molecular Weight (g/mol): 98.07 MDL Number: MFCD00064589 InChI Key: QAOWNCQODCNURD-UHFFFAOYSA-N Synonym: oil of vitriol,sulphuric acid,dihydrogen sulfate,mattling acid,battery acid,dipping acid,acide sulfurique,electrolyte acid,acidum sulfuricum,vitriol brown oil PubChem CID: 1118 ChEBI: CHEBI:26836 IUPAC Name: sulfuric acid SMILES: OS(O)(=O)=O
Sulfuric acid, ≥90% solution, J.T. Baker™
CAS: 7664-93-9 Molecular Formula: H2O4S Molecular Weight (g/mol): 98.07 MDL Number: MFCD00064589 InChI Key: QAOWNCQODCNURD-UHFFFAOYSA-N Synonym: oil of vitriol,sulphuric acid,dihydrogen sulfate,mattling acid,battery acid,dipping acid,acide sulfurique,electrolyte acid,acidum sulfuricum,vitriol brown oil PubChem CID: 1118 ChEBI: CHEBI:26836 IUPAC Name: sulfuric acid SMILES: OS(O)(=O)=O
Sulfuric Acid, 50% (v/v), Ricca Chemical
CAS: 7664-93-9 Molecular Formula: H2O4S Molecular Weight (g/mol): 98.07 MDL Number: MFCD00064589 InChI Key: QAOWNCQODCNURD-UHFFFAOYSA-N Synonym: oil of vitriol,sulphuric acid,dihydrogen sulfate,mattling acid,battery acid,dipping acid,acide sulfurique,electrolyte acid,acidum sulfuricum,vitriol brown oil PubChem CID: 1118 ChEBI: CHEBI:26836 IUPAC Name: sulfuric acid SMILES: OS(O)(=O)=O
Amine-Sulfuric Acid Reagent, for Sulfide Determination (Methylene Blue Method), Ricca Chemical
Sulfide Determination (Methylene Blue Method)
Sulfuric Acid, 20% (v/v), Reagent Grade, Ricca Chemical
CAS: 7664-93-9 Molecular Formula: H2O4S Molecular Weight (g/mol): 98.07 MDL Number: MFCD00064589 InChI Key: QAOWNCQODCNURD-UHFFFAOYSA-N PubChem CID: 1118 ChEBI: CHEBI:26836 IUPAC Name: sulfuric acid SMILES: OS(O)(=O)=O
Sulfuric Acid, Certified, 2.00N ±0.02N (1.0M), LabChem™
CAS: 7664-93-9 Molecular Formula: H2O4S Molecular Weight (g/mol): 98.07 MDL Number: MFCD00064589 InChI Key: QAOWNCQODCNURD-UHFFFAOYSA-N Synonym: oil of vitriol,sulphuric acid,dihydrogen sulfate,mattling acid,battery acid,dipping acid,acide sulfurique,electrolyte acid,acidum sulfuricum,vitriol brown oil PubChem CID: 1118 ChEBI: CHEBI:26836 IUPAC Name: sulfuric acid SMILES: OS(O)(=O)=O
Sulfuric Acid Concentrate (Certified) ACS, Fisher Chemical
CAS: 7664-93-9 Molecular Formula: H2O4S Molecular Weight (g/mol): 98.07 MDL Number: MFCD00064589 InChI Key: QAOWNCQODCNURD-UHFFFAOYSA-N Synonym: oil of vitriol,sulphuric acid,dihydrogen sulfate,mattling acid,battery acid,dipping acid,acide sulfurique,electrolyte acid,acidum sulfuricum,vitriol brown oil PubChem CID: 1118 ChEBI: CHEBI:26836 IUPAC Name: sulfuric acid SMILES: OS(O)(=O)=O
Sulfuric Acid ACS, 95 to 98%, MilliporeSigma™
CAS: 7664-93-9 Molecular Formula: H2O4S Molecular Weight (g/mol): 98.07 MDL Number: MFCD00064589 InChI Key: QAOWNCQODCNURD-UHFFFAOYSA-N Synonym: oil of vitriol,sulphuric acid,dihydrogen sulfate,mattling acid,battery acid,dipping acid,acide sulfurique,electrolyte acid,acidum sulfuricum,vitriol brown oil PubChem CID: 1118 ChEBI: CHEBI:26836 IUPAC Name: sulfuric acid SMILES: OS(O)(=O)=O
Sulfuric acid, 1.0N Standardized Solution
CAS: 7664-93-9 Molecular Formula: H2O4S Molecular Weight (g/mol): 98.07 MDL Number: MFCD00064589 InChI Key: QAOWNCQODCNURD-UHFFFAOYSA-N PubChem CID: 1118 ChEBI: CHEBI:26836 IUPAC Name: sulfuric acid SMILES: OS(O)(=O)=O
Sulfuric acid, 0.02N Standardized Solution
CAS: 7664-93-9 Molecular Formula: H2O4S Molecular Weight (g/mol): 98.07 MDL Number: MFCD00064589 InChI Key: QAOWNCQODCNURD-UHFFFAOYSA-N Synonym: oil of vitriol,sulphuric acid,dihydrogen sulfate,mattling acid,battery acid,dipping acid,acide sulfurique,electrolyte acid,acidum sulfuricum,vitriol brown oil PubChem CID: 1118 ChEBI: CHEBI:26836 IUPAC Name: sulfuric acid SMILES: OS(O)(=O)=O
Sulfuric acid, 0.1N Standardized Solution
CAS: 7664-93-9 Molecular Formula: H2O4S Molecular Weight (g/mol): 98.07 MDL Number: MFCD00064589 InChI Key: QAOWNCQODCNURD-UHFFFAOYSA-N PubChem CID: 1118 ChEBI: CHEBI:26836 IUPAC Name: sulfuric acid SMILES: OS(O)(=O)=O
Sulfuric acid, 0.2N Standardized Solution
CAS: 7664-93-9 Molecular Formula: H2O4S Molecular Weight (g/mol): 98.07 MDL Number: MFCD00064589 InChI Key: QAOWNCQODCNURD-UHFFFAOYSA-N IUPAC Name: sulfuric acid SMILES: OS(O)(=O)=O