accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3

CAS RN 7667-80-3

IUPAC Name: hexacontane
Synonyms: N-hexacontane
Molecular Weight (g/mol): 843.64
Molecular Formula: C60H122
InChi Key: OCWUCHKZAHTZAB-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results