accessibility menu, dialog, popup

UserName

Viewing profile as user:
KCH3(R)O(R)CH3(S)(S)CH3(S)(R)OHOO(R)

CAS RN 76676-34-1

IUPAC Name: potassium 3-[(1R,3aS,3bR,4R,9aR,9bS,11aS)-1-hydroxy-9a,11a-dimethyl-7-oxo-4-propyl-1H,2H,3H,3aH,3bH,4H,5H,7H,8H,9H,9aH,9bH,10H,11H,11aH-cyclopenta[a]phenanthren-1-yl]propanoate
Synonyms: potassium 3-[(1R,3aS,3bR,4R,9aR,9bS,11aS)-1-hydroxy-9a,11a-dimethyl-7-oxo-4-propyl-2H,3H,3aH,3bH,4H,5H,8H,9H,9bH,10H,11H-cyclopenta[a]phenanthren-1-yl]propanoate
Molecular Weight (g/mol): 440.67
Molecular Formula: C25H37KO4
InChi Key: HXJITUGMCJCKCE-UYOQDFFISA-M
SMILES: [K+].CCC[C@@H]1CC2=CC(=O)CC[C@]2(C)[C@H]2CC[C@@]3(C)[C@@H](CC[C@@]3(O)CCC([O-])=O)[C@H]12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results