accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 7669-54-7

IUPAC Name: [(2-nitrophenyl)sulfanyl]chlorane
Synonyms: [(2-nitrophenyl)sulfanyl]chlorane
Molecular Weight (g/mol): 189.61
Molecular Formula: C6H4ClNO2S
InChi Key: NTNKNFHIAFDCSJ-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=CC=C1SCl
Molecular Weight (g/mol) 
  • (4)
  • (2)
Percent Purity 
  • (2)
  • (4)
Quantity 
  • (2)
  • (1)
  • (1)
  • (2)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (4)
  • (4)
  • (2)
  • (2)
  • (4)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)