accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 767-63-5

IUPAC Name: 2,1,3-benzoxadiazol-4-amine
Synonyms: 2,1,3-benzoxadiazol-4-amine
Molecular Weight (g/mol): 135.13
Molecular Formula: C6H5N3O
InChi Key: IPCMVRZVNJHUHR-UHFFFAOYSA-N
SMILES: NC1=CC=CC2=NON=C12
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results