accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 76727-11-2

IUPAC Name: 6,13-diphenylpentacene
Synonyms: 6,13-diphenylpentacene
Molecular Weight (g/mol): 430.55
Molecular Formula: C34H22
InChi Key: PFCSVQKCECNEAZ-UHFFFAOYSA-N
SMILES: C1=CC=C(C=C1)C1=C2C=C3C=CC=CC3=CC2=C(C2=CC=CC=C2)C2=CC3=CC=CC=C3C=C12
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results