accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 767291-67-8

IUPAC Name: N1,N2-bis(3,3-dimethylbutyl)-N1,N2-dimethylcyclohexane-1,2-diamine
Synonyms: N1,N2-bis(3,3-dimethylbutyl)-N1,N2-dimethylcyclohexane-1,2-diamine
Molecular Weight (g/mol): 310.57
Molecular Formula: C20H42N2
InChi Key: VGCWVKVNKNXOGZ-UHFFFAOYNA-N
SMILES: CN(CCC(C)(C)C)C1CCCCC1N(C)CCC(C)(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results