accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3H3C(R)OHClNNNH3CCH3H3C(S)OHClNNN

CAS RN 76738-62-0

IUPAC Name: (3R)-1-(4-chlorophenyl)-4,4-dimethyl-2-(1H-1,2,4-triazol-1-yl)pentan-3-ol; (3S)-1-(4-chlorophenyl)-4,4-dimethyl-2-(1H-1,2,4-triazol-1-yl)pentan-3-ol
Synonyms: paclobutrazol
Molecular Weight (g/mol): 587.59
Molecular Formula: C30H40Cl2N6O2
InChi Key: GHGCUYKTGPFCTK-LZOIJMLOSA-N
SMILES: CC(C)(C)[C@@H](O)C(CC1=CC=C(Cl)C=C1)N1C=NC=N1.CC(C)(C)[C@H](O)C(CC1=CC=C(Cl)C=C1)N1C=NC=N1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results