accessibility menu, dialog, popup

UserName

Viewing profile as user:
HNSOHONHSNOO

CAS RN 7675-65-2

IUPAC Name: 3-(benzylsulfanyl)-2-{[(2-nitrophenyl)sulfanyl]amino}propanoic acid; N-cyclohexylcyclohexanamine
Synonyms: 3-(benzylsulfanyl)-2-{[(2-nitrophenyl)sulfanyl]amino}propanoic acid; dicha
Molecular Weight (g/mol): 545.76
Molecular Formula: C28H39N3O4S2
InChi Key: MAUIFYQGIWFDHJ-UHFFFAOYNA-N
SMILES: C1CCC(CC1)NC1CCCCC1.OC(=O)C(CSCC1=CC=CC=C1)NSC1=CC=CC=C1[N+]([O-])=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results