accessibility menu, dialog, popup

UserName

Viewing profile as user:
HO(R)OOHNOOCH3CH3CH3CH3

CAS RN 76757-90-9

IUPAC Name: methyl 2-{[(tert-butoxy)carbonyl]amino}-3-(4-hydroxyphenyl)propanoate
Synonyms: methyl 2-[(tert-butoxycarbonyl)amino]-3-(4-hydroxyphenyl)propanoate
Molecular Weight (g/mol): 295.34
Molecular Formula: C15H21NO5
InChi Key: NQIFXJSLCUJHBB-UHFFFAOYNA-N
SMILES: COC(=O)C(CC1=CC=C(O)C=C1)NC(=O)OC(C)(C)C
Quantity 
  • (1)
  • (1)
  • (2)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)