accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3C

CAS RN 768-49-0

IUPAC Name: (2-methylprop-1-en-1-yl)benzene
Synonyms: (2-methyl-1-propenyl)benzene
Molecular Weight (g/mol): 132.21
Molecular Formula: C10H12
InChi Key: BTOVVHWKPVSLBI-UHFFFAOYSA-N
SMILES: CC(C)=CC1=CC=CC=C1
Boiling Point 
  • (1)
  • (2)
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)