accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3CNH

CAS RN 768-52-5

IUPAC Name: N-(propan-2-yl)aniline
Synonyms: N-isopropylaniline
Molecular Weight (g/mol): 135.21
Molecular Formula: C9H13N
InChi Key: FRCFWPVMFJMNDP-UHFFFAOYSA-N
SMILES: CC(C)NC1=CC=CC=C1
Boiling Point 
  • (2)
  • (3)
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (2)
  • (3)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)