accessibility menu, dialog, popup

UserName

Viewing profile as user:
F

CAS RN 768-92-3

IUPAC Name: 1-fluoroadamantane
Synonyms: 1-fluoroadamantane
Molecular Weight (g/mol): 154.23
Molecular Formula: C10H15F
InChi Key: CPWSNJSGSXXVLD-UHFFFAOYSA-N
SMILES: FC12CC3CC(CC(C3)C1)C2
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)