accessibility menu, dialog, popup

UserName

Viewing profile as user:
OH

CAS RN 768-95-6

IUPAC Name: adamantan-1-ol
Synonyms: 1-hydroxyadamantane
Molecular Weight (g/mol): 152.24
Molecular Formula: C10H16O
InChi Key: VLLNJDMHDJRNFK-UHFFFAOYSA-N
SMILES: OC12CC3CC(CC(C3)C1)C2
Molecular Weight (g/mol) 
  • (2)
  • (5)
  • (3)
Percent Purity 
  • (2)
  • (8)
Quantity 
  • (1)
  • (1)
  • (3)
  • (2)
  • (2)
  • (1)
Physical Form 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (2)
  • (2)
  • (6)
  • (2)
  • (5)
  • (3)
  • (2)
  • (8)
  • (1)
  • (1)
  • (3)
  • (2)
  • (2)
  • (1)
  • (2)
  • (3)