CAS RN 7681-76-7
IUPAC Name:
(1-methyl-5-nitro-1H-imidazol-2-yl)methyl carbamate
Synonyms:
ronidazole
Molecular Weight (g/mol):
200.15
Molecular Formula:
C6H8N4O4
InChi Key:
PQFRTXSWDXZRRS-UHFFFAOYSA-N
SMILES:
CN1C(COC(N)=O)=NC=C1[N+]([O-])=O
1
–
1
of
1
results
1
Ronidazole, ≥95%, MilliporeSigma™ Supelco™
Ronidazole is a nitroimidazole compound, commonly used as antiprotozoal and antibacterial agent.