accessibility menu, dialog, popup

UserName

Viewing profile as user:
NaNaOSOONNSOOO

CAS RN 76836-02-7

IUPAC Name: disodium 2-[4-(2-sulfonatoethyl)piperazin-1-yl]ethane-1-sulfonate
Synonyms: pipes disodium salt disodium pipes
Molecular Weight (g/mol): 346.32
Molecular Formula: C8H16N2Na2O6S2
InChi Key: GMHSTJRPSVFLMT-UHFFFAOYSA-L
SMILES: [Na+].[Na+].[O-]S(=O)(=O)CCN1CCN(CCS([O-])(=O)=O)CC1
Percent Purity 
  • (1)
  • (3)
Physical Form 
  • (1)
  • (3)
Quantity 
  • (1)
  • (2)
  • (1)
pH 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (3)
  • (1)
  • (3)
  • (1)
  • (3)
  • (1)
  • (2)
  • (1)
  • (3)