accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNHOOOH3COSiCH3H3CCH3CH3H3C

CAS RN 76855-69-1

IUPAC Name: 3-{1-[(tert-butyldimethylsilyl)oxy]ethyl}-4-oxoazetidin-2-yl acetate
Synonyms: 3-{1-[(tert-butyldimethylsilyl)oxy]ethyl}-4-oxoazetidin-2-yl acetate
Molecular Weight (g/mol): 287.43
Molecular Formula: C13H25NO4Si
InChi Key: GWHDKFODLYVMQG-UHFFFAOYNA-N
SMILES: CC(O[Si](C)(C)C(C)(C)C)C1C(NC1=O)OC(C)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results