accessibility menu, dialog, popup

UserName

Viewing profile as user:
NOH

CAS RN 76876-70-5

IUPAC Name: 2-(1-benzylpiperidin-4-yl)ethan-1-ol
Synonyms: 2-(1-benzylpiperidin-4-yl)ethanol
Molecular Weight (g/mol): 219.33
Molecular Formula: C14H21NO
InChi Key: FBFPTWSKNJHCGT-UHFFFAOYSA-N
SMILES: OCCC1CCN(CC2=CC=CC=C2)CC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results